Order Entry
United States
ContactUsLinkComponent
Somso® Female Pelvis
Somso® Female Pelvis
Catalog # 470106-156
Supplier:  MARCUS SOMMER
undefined
Somso® Female Pelvis
Catalog # 470106-156
Supplier:  MARCUS SOMMER
Supplier Number:  MS 8/2

Specifications

  • Description:
    Female pelvis 2 Pt
  • Height:
    17 cm
  • Length:
    23 cm
  • Cat. No.:
    470106-156
  • Width:
    28 cm
  • Supplier no.:
    MS 8/2

Specifications

About this item

Somso® premium quality models.

  • 5 year warranty

As MS 8/1, but only separates into 2 parts. In SOMSO-Plast®.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.