ContactUsLinkComponent
Somso® Red Blood-Corpuscle
Catalog # 470321-038
Supplier:  MARCUS SOMMER
Somso® Red Blood-Corpuscle
Catalog # 470321-038
Supplier:  MARCUS SOMMER
Supplier Number:  HS20/1
Specifications
  • Description:
    Red blood cell
  • Cat. No.:
    470321-038
Specifications
About this item
Somso® premium quality models.
  • 5 year warranty
Enlarged approx. 11.000 times, in SOMSO-Plast®. In one piece.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.