Order Entry
United States
ContactUsLinkComponent
Somso® Half of the Brain Natural Cast
Somso® Half of the Brain Natural Cast
Catalog # 470313-144
Supplier:  MARCUS SOMMER
undefined
Somso® Half of the Brain Natural Cast
Catalog # 470313-144
Supplier:  MARCUS SOMMER
Supplier Number:  BS20/1

Specifications

  • Description:
    Half of the brain natural cast
  • Height:
    15 cm
  • Length:
    6 cm
  • Cat. No.:
    470313-144
  • Width:
    17 cm

Specifications

About this item

Somso® premium quality models.

  • 5 year warranty

Natural cast, in SOMSO-Plast®. Separates into 4 parts: frontal and parietal lobes, temporal and occipital lobes, medulla and cerebellum. On a base.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.