Order Entry
United States
ContactUsLinkComponent
Somso® Anatomical Structure of Pine Wood
Somso® Anatomical Structure of Pine Wood
Catalog # 470312-542
Supplier:  MARCUS SOMMER
undefined
Somso® Anatomical Structure of Pine Wood
Catalog # 470312-542
Supplier:  MARCUS SOMMER
Supplier Number:  BOS21

Specifications

  • Description:
    Structure of pinewood
  • Height:
    15 cm
  • Length:
    30 cm
  • Cat. No.:
    470312-542
  • Width:
    65 cm

Specifications

About this item

Somso® premium quality models.

  • 5 year warranty

Pinus silvestris, enlarged approx. 350 times, in SOMSO-Plast®. After Prof. Dr. W. Jung. The model shows the anatomical structure of pine wood in various sections: transverse, radial longitudinal and tangential longitudinal through the cambium, early wood, late wood and bark. Showing all the elements of the wood structure. In one piece. On a green base with description.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.