Order Entry
United States
ContactUsLinkComponent
Somso® Transparent Torso Model with Head
Somso® Transparent Torso Model with Head
Catalog # 470332-420
Supplier:  MARCUS SOMMER
undefined
Somso® Transparent Torso Model with Head
Catalog # 470332-420
Supplier:  MARCUS SOMMER
Supplier Number:  AS9/2

Specifications

  • Description:
    Transparent torso
  • Height:
    106 cm
  • Length:
    27 cm
  • Cat. No.:
    470332-420
  • Width:
    46 cm

Specifications

About this item

Somso® premium quality models.

  • Natural size
  • 5 year warranty

The transparent model shows the skeletal system in conjunction with the topography of the intestines. The cranial bones are not shown in order to expose the brain and the blood vessels supplying it. In one piece.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.