Order Entry
United States
ContactUsLinkComponent
Somso® Cervical Vertebral Column with Shoulder Girdle
Somso® Cervical Vertebral Column with Shoulder Girdle
Catalog # 470102-770
Supplier:  MARCUS SOMMER
undefined
Somso® Cervical Vertebral Column with Shoulder Girdle
Catalog # 470102-770
Supplier:  MARCUS SOMMER
Supplier Number:  QS 65/5

Specifications

  • Description:
    Cervical vertebrae
  • Height:
    59 cm
  • Length:
    30 cm
  • Cat. No.:
    470102-770
  • Width:
    39 cm
  • Supplier no.:
    QS 65/5

Specifications

About this item

Somso® premium quality models.

  • 5 year warranty

Natural size, in SOMSO-Plast®. Showing the cervical plexus and the brachial plexus. On a stand with green base.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.