Order Entry
United States
ContactUsLinkComponent
Somso® Skeleton Transparent
Somso® Skeleton Transparent
Catalog # 470102-554
Supplier:  MARCUS SOMMER
undefined
Somso® Skeleton Transparent
Catalog # 470102-554
Supplier:  MARCUS SOMMER
Supplier Number:  QS 10/12T

Specifications

  • Description:
    Skeleton transparent
  • Height:
    180 cm
  • Length:
    55 cm
  • Cat. No.:
    470102-554
  • Width:
    55 cm
  • Supplier no.:
    QS 10/12T

Specifications

About this item

Somso® premium quality models.

  • 5 year warranty

Natural cast of the bonesof a male adult, in SOMSO-Plast®. Showing life-size all the anatomical details of the bone structure. Skull with removable vault and mandible. Joints mounted and movable, upper and lower extremities can be removed. Left and right feet can be removed from the leg. With flexible vertebral column. Mounted on a stand. with rollers.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.