Order Entry
United States
ContactUsLinkComponent
Somso® Fertilization and Development of the Human Ovum Up To the 3rd Month
Somso® Fertilization and Development of the Human Ovum Up To the 3rd Month
Catalog # 470121-954
Supplier:  MARCUS SOMMER
undefined
Somso® Fertilization and Development of the Human Ovum Up To the 3rd Month
Catalog # 470121-954
Supplier:  MARCUS SOMMER
Supplier Number:  MS 15

Specifications

  • Description:
    Human ovum up to the 3rd month
  • Height:
    50 cm
  • Length:
    11 cm
  • Cat. No.:
    470121-954
  • Width:
    58 cm
  • Supplier no.:
    MS 15

Specifications

About this item

Somso® premium quality models.

  • 5 year warranty

Shown by 16 different models, in SOMSO-Plast®. Collection in a display case with removable transparent cover.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.