Order Entry
United States
ContactUsLinkComponent
Somso® Thoracic Vertebra (Th II) with Spinal Cord
Somso® Thoracic Vertebra (Th II) with Spinal Cord
Catalog # 470313-146
Supplier:  MARCUS SOMMER
undefined
Somso® Thoracic Vertebra (Th II) with Spinal Cord
Catalog # 470313-146
Supplier:  MARCUS SOMMER
Supplier Number:  BS28/1

Specifications

  • Description:
    Thoracic vertebra with spinal cord
  • Height:
    14 cm
  • Length:
    16 cm
  • Cat. No.:
    470313-146
  • Width:
    14 cm

Specifications

About this item

Somso® premium quality models.

  • 5 year warranty

Natural size, in SOMSO-Plast®. Spinal nerves, spinal ganglion and spinal cord are shown in cross-section. In one piece. On a stand with green base.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.