Order Entry
United States
Orders LinkContactUsLinkComponent
 

Somso® Primrose

Primrose, 33 cm, 12 cm, 42 cm
Catalog # 470312-450
Supplier:   MARCUS SOMMER
 :  BOS15/4
Somso® Primrose
Catalog # 470312-450
Supplier:  MARCUS SOMMER
Somso® Primrose
Somso® Primrose

Primrose, 33 cm, 12 cm, 42 cm

Price
$770.00
Log in for availability
Each
Product Details & Documents

Somso® premium quality models.

  • 5 year warranty

Primula officinalis (cowslip), in SOMSO-Plast®. After Prof. Dr. W. Weber. Median section through two heterostyled smelling primrose flowers, enlarged approx. 13 times. The relief presentation highlights the heterostyly to facilitate cross fertilization. In one piece. On a stand and green base with explanatory note.

 : Items on this page are not stocked and may have an extended time for delivery.

Specifications
  • Description:
    Primrose
  • Height:
    42 cm
  • Length:
    12 cm
  • Width:
    33 cm