Orders LinkContactUsLinkComponent
Eisco® Model Lymph Node
Eisco® Model Lymph Node
Lymph nodes model
Catalog # 470356-230
Supplier:   Wards
Supplier #:  AM0306
Eisco® Model Lymph Node
Catalog # 470356-230
Supplier:  Wards
Eisco® Model Lymph Node
Eisco® Model Lymph Node
Lymph nodes model
Price
$336.45
Log in for availability
Each
Product Details & Documents
An easy to understand lymph node.
  • Mounted on base
  • Includes key card
  • Enlarged size
Model shows incoming lymph vessel, germinal centre, sinus, outgoing lymph vessel, capsule, Trabecula, incoming blood supply artery, and vein for outgoing blood. Lymph nodes vary in diameter from 1 to 25 mm, although they can swell during infection or illness. They are covered in a fibrous capsule, they contain sinuses, where many scavenging white blood cells, called macrophages, ingest bacteria as well as other foreign matter and debris.
Specifications
  • Description:
    Lymph nodes model