Order Entry
United States
ContactUsLinkComponent
ProMTag Peptide Universal Protein Cleanup Kit, Impact Proteomics
ProMTag Peptide Universal Protein Cleanup Kit, Impact Proteomics
Catalog # MSPP-PEP-1001
CAS Number:  
undefined
ProMTag Peptide Universal Protein Cleanup Kit, Impact Proteomics
Catalog # MSPP-PEP-1001
Supplier Number:  PEP-1001
CAS Number:  

Specifications

  • Description:
    ProMTag Peptide Universal Protein Cleanup Kit
  • Size:
    1 kit
  • Cat. No.:
    MSPP-PEP-1001
  • Details:
    Protein amount per sample 1 – 40 µg
  • Sufficient for:
    8 samples

Specifications

About this item

The ProMTag Peptide Universal Protein Cleanup Kit produces high-quality peptide samples ready for mass spectrometry analysis.

  • Compatible with most starting samples including cultured cells, tissues, biological fluids, and pre-made lysates so long as they do not contain buffers with primary amines (such as TRIS).
  • All reagents and plasticware for 8 sample preparations are included.
  • Includes Impact Proteomics' MT-Trypsin. The user does not need to provide an enzyme.
  • Requires only standard laboratory equipment.

The ProMTag Peptide Universal Protein Cleanup Kit utilizes the reversible chemistry of ProMTag to produce high-quality peptide samples for mass spectrometry analysis in a simple, fast workflow with minimal hands-on time. Works with any starting sample for rapid, high yield extraction and cleanup of protein samples, and digestion of proteins into peptides. Each kit contains reagents for 8 sample preparations.