Polygun™ Adhesive Applicators, 3M™
Supplier: 3M
Certificates
About this item
Three models of applicators are available for dispensing 3M Jet-melt™ adhesives (see 12778-018 series).
The Polygun™ II portable, self-contained, pneumatic applicator delivers adhesive at a rate of up to 3.4kg (7.5lbs.) per hour. Comes with speedloader cartridge feed and bench stand.
The all-electric Polygun™ TC with 0.23cm (0.09") tip is rugged, reliable, and compact. It weighs only 238g (10oz.) and delivers 1.6kg (3.5lbs.) of adhesive per hour.
For low melting adhesives, the Polygun™ LT with 0.23cm (0.09") tip operates at just 129°C (265°F).
Frequently Bought Together