Mechanical Wave Kit Parts and Accessories
Essential replacement parts for your mechanical wave investigations.
- 40 grams of replacement blue sand for chladni plates
- 0.5 mm thick×1 meter long string for demonstrating standing waves
- Replacement spring has 0.75 mm thick wire, and measures 30 mm diameter×60 mm length (compressed)
Strings snap, springs deform, and sand gets swept into the garbage can. Keep your mechanical wave demonstrations ready to go with these high-quality replacement parts.
Frequently Bought Together