Mechanical Vibration Generator
Catalog # 470352-704
Supplier: GSC International
Specifications
- Description:Mechanical vibration generator
- Cat. No.:470352-704
- Amps:max. 1 A
Specifications
About this item
Generate physical oscillations in the lab for studying the properties of mechanical waves.
- Requires a variable frequency sine wave signal generator
- Electrical input by 4 mm banana plugs, 1A max
Looking for a hands-on way to explore oscillation and resonance in your STEM classroom. This is the perfect companion to a variable frequency sine wave signal generator. Designed for hassle-free wave demonstrations, this compact device generates a steady supply of consistent waves, allowing students to dive deep into the fascinating world of resonance and vibration.
Simply pair it with a sine wave signal generator, and you'll have everything needed to captivate students and bring wave theory to life. No complicated setup, no fuss - just plug in and start exploring the mesmerizing patterns that unfold before your eyes.