Digital Ammeter
Product Details & Documents
Perfect for a wide range of electrical projects and experiments.
- Easy-to-read display
- Powered by 9 V battery, included
Stop struggling with outdated analog meters or complex setups. This compact, battery-powered, digital device provides accurate readings from 0.00 to 19.99 A, making it perfect for a wide range of science, education, and DIY projects. With its easy-to-read display, you can quickly check the amps flowing through your circuits without any guesswork.
Simply connect it in series, and the digital ammeter will do the rest, powered by a simple 9 V battery. Whether you're a student, educator, or electronics enthusiast, this reliable ammeter is an essential tool to have on hand. Unlock the secrets of electricity, explore STEM concepts, or troubleshoot your latest creation - the digital ammeter puts the power of precise current measurement right at your fingertips.
Specifications
- Description:Digital ammeter, 0 - 19.99 A