Orders LinkContactUsLinkComponent
DC Motor
DC Motor
DC motor with leads, 1.5 – 3 V
  470352-706
Supplier #:  4-280
DC Motor
  470352-706
DC Motor
DC Motor
DC motor with leads, 1.5 – 3 V
$4.49
Each
Don't let the size fool you - this motor packs a punch.
  • 2 connecting leads in red and black
  • A useful for many STEM activities
Looking for a powerful little motor to power your next STEM project. This compact workhorse operates on just 1.5 to 3 volts, yet packs a punch with an impressive 11,000 rpm. Whether you're building a robot, a model car, or experimenting with electrical circuits, this motor is a versatile and reliable component. With its simple red and black connecting leads, it's easy to integrate into your designs. It's perfect for all kinds of hands-on science and engineering activities, from physics demonstrations to prototyping innovative new inventions.
  • Voltage:
    1.5 – 3 V
  • Description:
    DC motor with leads