Sharps Disposal Containers with Horizontal-Drop Opening Lids, Covidien
Supplier: Cardinal Health
Certificates
About this item
Clear lid with large horizontal opening accommodates larger syringes, allowing one-hand, one-step, no-touch sharps depositing
Anti-spill inner flap on the lid impedes hand entry. Container base has hand grips for easy transport. Containers lock for final disposal. Plastics and pigments are safe to autoclave or incinerate. Containers can be placed on countertops or mounted on a wall.
Certifications: Containers meet or exceed JCAHO, OSHA, and NIOSH/CDC standards and requirements.
Frequently Bought Together