LidLocks™ Microcentrifuge Tube Locks, Sorenson BioScience
14229-941
: Avantor
- Color:Blue
- Cat. No.:14229-941
- Supplier no.:11870
The unique design of LidLocks™ tube locks provides extra security against inadvertent cap openings on critical samples
Ideal for shipping samples. Fit 1.5mL and 1.7mL microcentrifuge tubes. Certified RNase-, DNase-, and pyrogen-free.