ContactUsLinkComponent
Silver Shield®/4H® Sleeves, Honeywell Safety
Silver Shield®/4H® Sleeves, Honeywell Safety
Catalog # 11000-630
undefined
Silver Shield®/4H® Sleeves, Honeywell Safety
Catalog # 11000-630
Supplier Number:  SSS
Specifications
  • Garment application:
    Chemical protection,
    Laboratory
  • Description:
    Silver Shield®/4H® Sleeves
  • Cat. No.:
    11000-630
  • Supplier no.:
    SSS
Specifications
About this item
Sleeves feature a five-layer fabric compacted into 2.7mil
Laminating a high-energy polymer (EVOH) with a low-energy polymer (PE) creates a fabric impervious to at least 90% of all organic chemicals for four hours at 35°C (95°F). Each layer acts as a barrier against various epoxies and industrial chemicals. Can be used with Silver Shield®/4H® gloves and/or apron. Sleeves provide added protection for the upper arms and shoulders. Elastic bands at the shoulder and wrist ensure a snug fit. One size fits all.