Corning® 3-Hole Delivery Cap, High Temperature, Corning
Catalog # 62890-448
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Cap Size:GL45
- Cat. No.:62890-448
- Supplier no.:1395-45DC
Specifications
About this item
Red polybutylene terephthalate (PBT) GL45 screw cap has three 6.4mm (1/4") holes at top for use with 3.2mm (1/8") O.D
tubing. Caps have fluoropolymer resin liner for efficient sealing and may be heat sterilized at up to 180°C (356°F).