Orders LinkContactUsLinkComponent
Polypropylene Screw Caps, BrandTech
Polypropylene Screw Caps, BrandTech
Supplier:   Brandtech
Polypropylene Screw Caps, BrandTech
Supplier:  Brandtech
Certificates
Polypropylene Screw Caps, BrandTech
Polypropylene Screw Caps, BrandTech
3 Options Available Below
Product Details & Documents
Translucent screw caps are for use with BrandTech polypropylene and low-density polyethylene bottles.
  • Autoclavable translucent polypropylene
  • Fits VITLAB and many other bottles with standard thread
Packaging: Bulk packed.
Product Family Options
Product Information
  • Cap Size
    Availability
    Price
  • GL18
    Log in for availability
    Log in for availability
    Pack of 24
    Pack of 24 - $57.17
    $57.17
    New Customer Promotional Price
    Pack of 24
       $57.17
    Minimum order quantity is 1
    Supplier Number: 83310
  • GL32
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $29.23
    $29.23
    New Customer Promotional Price
    Pack of 12
       $29.23
    Minimum order quantity is 1
    Supplier Number: 83312
  • GL45
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $39.31
    $39.31
    New Customer Promotional Price
    Pack of 12
       $39.31
    Minimum order quantity is 1
    Supplier Number: 83313