Monkeypox Virus (MPXV) Recombinant B6R Protein (His Tag) (From HEK293 Cells), Biotinylated
77447-846
:
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Monkeypox virus
- Size:20 µg
- Tag sequence:His Tag
- Storage Conditions:Samples are stable for up to twelve months from date of receipt at −20 to −80 ℃. Store it under sterile conditions at −20 to −80 ℃. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method
- Reconstitution Instructions:A hardcopy of the datasheet with reconstitution instructions is sent together with the product. Please refer to it for detailed information.
- Endotoxin-free:No
- Carrier-Free:Yes
- Animal-Free:No
- Protein Synonyms:Monkeypox Virus B6R
- Protein/Peptide Name:MPXV-B6R
- Purity:>85% as determined by SDS-PAGE.
- Molecular Weight:The recombinant Monkeypox Virus (MPXV) B6R Protein consists of 273 amino acids and predicts a molecular mass of 30.52 kDa. It migrates as an approximately 41.04 kDa band in SDS-PAGE under reducing conditions.
- Shipping Temperature:Ambient
- Cat. No.:77447-846
A DNA sequence encoding the Monkeypox Virus (MPXV) B6R Protein (QNP13760.1) (Tyr18-His279) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.