Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant CD200R1/CD200R Protein (His Tag) (From HEK293 Cells)
  77447-038
 :  
Human Recombinant CD200R1/CD200R Protein (His Tag) (From HEK293 Cells)
  77447-038
 :  11218-H08H-1
 :  

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    1 mg
  • Tag sequence:
    His Tag
  • Storage Conditions:
    Samples are stable for up to twelve months from date of receipt at −20 to −80 ℃. Store it under sterile conditions at −20 to −80 ℃. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Measured by its binding ability in a functional ELISA
    . Immobilized human CD200R1 at 1 μg/ml (100 μl/well) can bind human CD200 with a linear ra nge of 0.12 - 16 ng/ml.
  • Reconstitution Instructions:
    A hardcopy of the datasheet with reconstitution instructions is sent together with the product. Please refer to it for detailed information.
  • Endotoxin-free:
    No
  • Carrier-Free:
    Yes
  • Animal-Free:
    No
  • Protein Synonyms:
    CD200R,HCRTR2,MOX2R,OX2R
  • Protein/Peptide Name:
    CD200R1
  • Purity:
    ≥98% as determined by SDS-PAGE. ≥95% as determined by SEC-HPLC.
  • Molecular Weight:
    The secreted recombinant human CD200R1 comprises 251 amino acids after removal of the signal peptide and has a predicted molecular mass of 28.2 kDa. As a result of glycosylation, rh CD200R1 migrates as an approximately 50-60 kDa band in SDS-PAGE under reducing conditions.
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    77447-038

 

 

A DNA sequence encoding the human CD200R1 (AAI43394.1) extracellular domain (Met 1-Leu 266) was fused with a polyhistidine tag at the C-terminus.