Human Recombinant ANXA5/Annexin Ⅴ/Annexin A5 Protein (From E. coli), Biotinylated
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human ANXA5 (P08758) (Met1-Asp320) was expressed and purified. The purified protein was biotinylated in vitro.
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:Samples are stable for up to twelve months from date of receipt at −20 to −80 ℃. Store it under sterile conditions at −20 to −80 ℃. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
- Reconstitution Instructions:A hardcopy of the datasheet with reconstitution instructions is sent together with the product. Please refer to it for detailed information.
- Endotoxin-free:No
- Carrier-Free:Yes
- Animal-Free:No
- Protein Synonyms:ANX5,ENX2,HEL-S-7,PP4,RPRGL3
- Protein/Peptide Name:ANXA5
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant human ANXA5 consists of 320 amino acids and predicts a molecular mass of 35.9 kDa.
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together