Human Recombinant HER2/ErbB2 Protein (ECD, domain I) (His Tag) (From HEK293 Cells), Biotinylated
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human ERBB2 (AAA75493.1) (Met1-Cys195) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag
- Storage Conditions:Samples are stable for up to twelve months from date of receipt at −20 to −80 ℃. Store it under sterile conditions at −20 to −80 ℃. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method
- Reconstitution Instructions:A hardcopy of the datasheet with reconstitution instructions is sent together with the product. Please refer to it for detailed information.
- Endotoxin-free:No
- Carrier-Free:Yes
- Animal-Free:No
- Protein Synonyms:NEU|MLN19|HERVW|TKR1|HERV7Q|MLN 19|CD340|ERVWE1|HER2|HERV-W-ENV|ENV|HER-2/neu|HER-2|ENVW|HERVWENV|HERV-7q|NGL
- Protein/Peptide Name:ERBB2
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:The recombinant human ERBB2 consists of 184 amino acids and predicts a molecular mass of 20.9 kDa.
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together