ContactUsLinkComponent
Liebig Condenser, Chemglass
Catalog # 80067-398
Supplier:  Chemglass
Small Business Enterprise
undefined
Liebig Condenser, Chemglass
Catalog # 80067-398
Supplier:  Chemglass
Supplier Number:  CG-1218-A-20
Small Business Enterprise
Specifications
  • Description:
    Liebig Condenser
  • Cat. No.:
    80067-398
  • Supplier no.:
    CG-1218-A-20
Specifications
About this item
Small scale Liebig condenser with a [ST] outer joint at the top and a lower inner drip tip joint
Hose connections have an O.D. of 10mm at the middle serration. [ST] joint: 14/20.