Reflux Condenser, Chemglass
Catalog # 80067-364
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:Reflux Condenser
- Cat. No.:80067-364
- Supplier no.:CG-1217-20
Specifications
About this item
Compact, high efficiency reflux condenser employs a double wall thimble-shaped bulb sealed into the outer jacket
Serrated hose connections for water connection extend through the outer bulb and have an O.D. of 10mm in the middle serration. While the overall height of this condenser is only 210mm, it has the efficiency of a standard 400mm Liebig or West condenser. [ST] joint: 24/40.