CPE Booties, InterWorld
37001-074
- Shoe size:Universal
- Garment application:Laboratory
- Color:White
- Cat. No.:37001-074
- Supplier no.:10135-108-02
Water-resistant booties made from heavyweight and practically impervious compressed polyethylene (CPE) film, excellent for containing moisture and dirt from street shoes
CPE products guaranteed to meet 4.0 AQL level II sampling plan per MIL-STD-105E for: broken seams, broken elastic band, burnt/dirty joints, incomplete heat sealed joints, dirty material, physical dimensions non-compliance.
Resistant to chemical corrosion and heavily textured for better grip. Feature offset hole opening and seamless bottom. Dimensions: 40.6Lx22.9Hcm (16x9").