ContactUsLinkComponent
KLEENGUARD® A20 Breathable Particle Protection Boot Covers, KIMBERLY-CLARK PROFESSIONAL®
KLEENGUARD® A20 Breathable Particle Protection Boot Covers, KIMBERLY-CLARK PROFESSIONAL®
Catalog # 10837-310
Supplier:  Ansell Healthcare
   Waste Reducing
undefined
KLEENGUARD® A20 Breathable Particle Protection Boot Covers, KIMBERLY-CLARK PROFESSIONAL®
Catalog # 10837-310
Supplier:  Ansell Healthcare
Supplier Number:  36880
   Waste Reducing
Specifications
  • Shoe size:
    Universal
  • Garment application:
    Chemical protection
  • Environmentally Preferable:
    Waste Reducing
  • Cat. No.:
    10837-310
  • Supplier no.:
    36880
Specifications
About this item
Three-layer, MicroForce™ Barrier SMS fabric helps to control and filter dirt for process protection
Generous size covers work shoes. Boot covers have an elastic top for secure protection.