Rat OX (Orexin) ELISA Kit
Catalog # 77527-004
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Competitive
- Host:Rat
- Target protein:OX (Orexin)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:2 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:15.63 - 1000 pg/ml
- Storage Temperature:2…8 °C
- Application:Neuro science
- Sensitivity:4.53 pg/ml
- Regulatory Status:RUO
- Cat. No.:77527-004