Tissue Culture Tray, Mitchell Plastics™
Catalog # 29186-500
Supplier: Avantor
Specifications
- Environmentally Preferable:
- Cat. No.:29186-500
- W×D×H:19 x 40 x 5.1 cm (71/2 x 153/4 x 2")
- Supplier no.:ML-2600
Specifications
About this item
Holds tissue culture dishes and flasks during storage, transport, or incubation
Supports fourteen 100mm dishes or four 250mL flasks. Six slots on the bottom of the tray allow airflow when used in an incubator. A recessed bottom plate makes trays stackable.