Human Recombinant SEMA4D/Semaphorin 4D Protein (ECD, Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human SEMA4D (AAH54500.1) (Met1-Arg734) was expressed with the Fc region of human IgG1 at the C-terminus.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:ECD, Fc Tag
- Storage Conditions:Samples are stable for up to twelve months from date of receipt at −20...−80 °C. Store it under sterile conditions at −20...−80 °C. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method
- Reconstitution Instructions:A hardcopy of the datasheet with reconstitution instructions is sent together with the product. Please refer to it for detailed information.
- Endotoxin-free:No
- Carrier-Free:Yes
- Animal-Free:No
- Protein Synonyms:C9orf164|M-sema-G|SEMAJ|COLL4|CD100|coll-4
- Protein/Peptide Name:SEMA4D
- Purity:>90 % as determined by SDS-PAGE.
- Molecular Weight:The recombinant human SEMA4D consists of 951 amino acids and predicts a molecular mass of 105.95 kDa.
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together