Human Recombinant CD55/DAF Protein (Fc Tag) (from HEK293 Cells)
Catalog # 77446-294
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:1 mg
- Tag sequence:Fc Tag
- Storage Conditions:Samples are stable for up to twelve months from date of receipt at −20...−80 °C. Store it under sterile conditions at −20...−80 °C. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured by its ability to bind human CD97-His (Cat:11280-H08H) in a functional ELISA.
- Reconstitution Instructions:A hardcopy of the datasheet with reconstitution instructions is sent together with the product. Please refer to it for detailed information.
- Endotoxin-free:No
- Carrier-Free:Yes
- Animal-Free:No
- Protein Synonyms:CR|DAF|CROM|TC
- Protein/Peptide Name:CD55
- Purity:≥90 % as determined by SDS-PAGE. ≥95 % as determined by SEC-HPLC.
- Molecular Weight:The recombinant human CD55/Fc is a disulfide-linked homodimeric protein. The reduced monomer consists of 557 amino acids and has a predicted molecular mass of 61.7 kDa. In SDS-PAGE under reducing conditions, the apparent molecular mass of rhCD55/Fc monomer is approximately 95-105 kDa due to glycosylation.
- Shipping Temperature:Ambient
- Cat. No.:77446-294
Specifications
About this item
A DNA sequence encoding the human CD55 precursor (NP_000565.1) (Met 1-Ser 353) was expressed with the C-terminal fused Fc region of human IgG1.