ContactUsLinkComponent
Synthware Kjeldahl Trap Adapters, Kemtech America
Synthware Kjeldahl Trap Adapters, Kemtech America
Catalog # 60000-712
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Kjeldahl Trap Adapters, Kemtech America
Catalog # 60000-712
Supplier:  Kemtech America
Supplier Number:  A172440
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • [ST] joint:
    24/40
  • Cat. No.:
    60000-712
  • Supplier no.:
    A172440
Specifications
About this item
Kjeldahl trap adapters feature two [ST] 24/40 joints.
  • Used to catch splashes
  • With two joints
The span between the centers of joints is approximately 200 mm.