Orders LinkContactUsLinkComponent
Synthware Bent Adapters, 75°
Synthware Bent Adapters, 75°
Supplier:   Kemtech America
Synthware Bent Adapters, 75°
Supplier:  Kemtech America
Certificates
Synthware Bent Adapters, 75°
Synthware Bent Adapters, 75°
3 Options Available Below
Product Details & Documents
Synthware 75° bent adapters.
  • 75° angle bend
  • With inner joints
Adapters feature inner [ST] joints at each end.
Product Family Options
Product Information
  • [ST] joint no.
    Availability
    Price
  • 14/20
    Log in for availability
    Log in for availability
    Each
    Each - $36.62
    $36.62
    New Customer Promotional Price
    Each
       $36.62
    Minimum order quantity is 1
    Supplier Number: A151420
  • 19/22
    Log in for availability
    Log in for availability
    Each
    Each - $37.41
    $37.41
    New Customer Promotional Price
    Each
       $37.41
    Minimum order quantity is 1
    Supplier Number: A151922
  • 24/40
    Log in for availability
    Log in for availability
    Each
    Each - $60.22
    $60.22
    New Customer Promotional Price
    Each
       $60.22
    Supplier Number: A152440