Synthware Distillation Head Adapter, Kemtech America
Synthware Distillation Head Adapter
Catalog # 60000-642
Supplier: Kemtech America
Supplier #: A542440
Jump to:
Product Details & Documents
Synthware distillation head adapter is vacuum jacketed.
- [ST] 10/18 top joint
- Accommodates thermometer
- Thermometer not included
The [ST] 10/18 top joint accommodates a 76mm immersion thermometer. Other [ST] joints are 24/40.
: Thermometer is not supplied; contact your Avantor representative for more information.
Specifications
- Description:Synthware Distillation Head Adapter