Rat COL9a1 (Collagen Type IX Alpha 1) ELISA Kit
Catalog # 77521-374
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Rat
- Target protein:COL9a1 (Collagen Type IX Alpha 1)
- Size:96 wells
- Sample Type:Tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:1.57 - 100 ng/ml
- Storage Temperature:2…8 °C
- Application:Metabolic pathway, Ophthalmology and Otorhinolaryngology, Developmental science, Nutrition metabolism, Rheumatology, Autoimmunity
- Sensitivity:0.58 ng/ml
- Regulatory Status:RUO
- Cat. No.:77521-374