Human APLP1 (Amyloid Beta Precursor Like Protein 1) ELISA Kit
Catalog # 77518-242
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:APLP1 (Amyloid Beta Precursor Like Protein 1)
- Size:96 wells
- Sample Type:Tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.32 - 20 ng/ml
- Storage Temperature:2…8 °C
- Application:Metabolic pathway
- Sensitivity:0.118 ng/ml
- Regulatory Status:RUO
- Cat. No.:77518-242