Human ASAH1 (N-Acylsphingosine Amidohydrolase 1) ELISA Kit
Catalog # 77515-350
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:ASAH1 (N-Acylsphingosine Amidohydrolase 1)
- Size:96 wells
- Sample Type:Tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:1.57 - 100 ng/ml
- Storage Temperature:2…8 °C
- Application:Enzyme and Kinase
- Sensitivity:0.63 ng/ml
- Regulatory Status:RUO
- Cat. No.:77515-350