Human TNFb (Tumor Necrosis Factor Beta) ELISA Kit
Catalog # 77514-054
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:TNFb (Tumor Necrosis Factor Beta)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:15.63 - 1000 pg/ml
- Storage Temperature:2…8 °C
- Application:Cytokine, Tumor immunity, Infection immunity
- Sensitivity:23.6 pg/ml
- Regulatory Status:RUO
- Cat. No.:77514-054