Human CRP (C Reactive Protein) ELISA Kit
77511-522
:
- Assay Type:Sandwich
- Host:Human
- Target protein:CRP (C Reactive Protein)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, urine, cerebrospinal fluid, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:62.5 - 4000 pg/ml
- Storage Temperature:2…8 °C
- Application:Tumor immunity, Infection immunity, Cardiovascular biology, Rheumatology
- Sensitivity:26.1 pg/ml
- Regulatory Status:RUO
- Cat. No.:77511-522