Horse FGF9 (Fibroblast Growth Factor 9) ELISA Kit
77527-072
:
- Assay Type:Sandwich
- Host:Horse
- Target protein:FGF9 (Fibroblast Growth Factor 9)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:62.5 - 4000 pg/ml
- Storage Temperature:2…8 °C
- Application:Cytokine, Tumor immunity, Infection immunity
- Sensitivity:23.2 pg/ml
- Regulatory Status:RUO
- Cat. No.:77527-072