E1S (Estrone Sulfate) ELISA Kit
Catalog # 77525-128
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Competitive
- Target protein:E1S (Estrone Sulfate)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:2 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:1.57 - 100 ng/ml
- Storage Temperature:2…8 °C
- Application:Reproductive science
- Sensitivity:0.51 ng/ml
- Regulatory Status:RUO
- Cat. No.:77525-128
Specifications
About this item
- Host: General