Mouse MBP (Myelin Basic Protein) ELISA Kit
Catalog # 77515-972
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Mouse
- Target protein:MBP (Myelin Basic Protein)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.32 - 20 ng/ml
- Storage Temperature:2…8 °C
- Application:Infection immunity, Rheumatology, Dermatology
- Sensitivity:0.114 ng/ml
- Regulatory Status:RUO
- Cat. No.:77515-972