Human DNM1 (Dynamin 1) ELISA Kit
77514-844
:
- Assay Type:Sandwich
- Host:Human
- Target protein:DNM1 (Dynamin 1)
- Size:96 wells
- Sample Type:Tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.32 - 20 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, Metabolic pathway, Developmental science
- Sensitivity:0.116 ng/ml
- Regulatory Status:RUO
- Cat. No.:77514-844