Tech-Med® Medicine Cups, DUKAL
Catalog # 76235-140
Supplier: Dukal
Specifications
- Capacity:2 oz.
- Description:Medicine Cup
- Material:Stainless steel
- Cat. No.:76235-140
Specifications
About this item
Made of the high grade stainless steel (18/8, 304).
- Crafted without seams which prevents bacteria growth and allows for easy cleaning
- Autoclavable
- Graduated inside