Sheep VCAM1 (Vascular Cell Adhesion Molecule 1) ELISA Kit
Catalog # 77528-234
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Sheep
- Target protein:VCAM1 (Vascular Cell Adhesion Molecule 1)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:78.13 - 5000 pg/ml
- Storage Temperature:2…8 °C
- Application:CD and Adhesion molecule, Tumor immunity, Infection immunity
- Sensitivity:30 pg/ml
- Regulatory Status:RUO
- Cat. No.:77528-234