Order Entry
United States
ContactUsLinkComponent
Human EDC1 (Enhancer Of mRNA Decapping Protein 1) ELISA Kit
Human EDC1 (Enhancer Of mRNA Decapping Protein 1) ELISA Kit
Catalog # 77523-482
Supplier:  AFG Bioscience
CAS Number:  
undefined
Human EDC1 (Enhancer Of mRNA Decapping Protein 1) ELISA Kit
Catalog # 77523-482
Supplier:  AFG Bioscience
Supplier Number:  EK246533
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Host:
    Human
  • Target protein:
    EDC1 (Enhancer Of mRNA Decapping Protein 1)
  • Size:
    96 wells
  • Sample Type:
    Tissue homogenates and other biological fluids
  • Time to Results:
    3.5 h
  • Shelf Life:
    6 months (4 °C); 12 months (–20 °C)
  • Detection Range:
    0.16 - 10 ng/ml
  • Storage Temperature:
    2…8 °C
  • Application:
    Human gene deletion
  • Sensitivity:
    0.064 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    77523-482

Specifications

About this item