Mouse PDGFA (Platelet Derived Growth Factor Subunit A) ELISA Kit
77519-042
:
- Assay Type:Sandwich
- Host:Mouse
- Target protein:PDGFA (Platelet Derived Growth Factor Subunit A)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:78.13 - 5000 pg/ml
- Storage Temperature:2…8 °C
- Application:Cytokine, Tumor immunity, Infection immunity, Bone metabolism
- Sensitivity:32 pg/ml
- Regulatory Status:RUO
- Cat. No.:77519-042